Viscosity (cP)
Fluid
Custom
Water
Ethylene Glycol/Water 50/50
Propylene Glycol/Water 50/50
HFE 7200
Density
g/cm3
lb/ft3
Flow
lpm
gpm
Number of 90° elbows
Tube ID
millimeters
inches
Tube length
meters
feet
Application pressure loss
bar
psi
Vertical rise (meters)
Fill out the form for pressure loss results.
Additional notes:
The actual calculations are made using formulas based on smooth tubing and are approximations.
Actual pressure loss may vary based on the tubing, fittings, accessories, fluid, and temperature.
The plumbing and total pressure loss represents the differential pressure between the supply and return of the temperature control unit (TCU) and does not include any pressure losses internal to the TCU that would also be reflected in the pressure gauge reading.